Learn More
Sigma Aldrich Fine Chemicals Biosciences Peptone Hy-Soy T | 250g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P6463250G
Encompass
Peptone Hy-SoyT has been used as a component of malt extract peptone agar (MEPA) to cultivate Trichoderma viride. It is also useful in media where high clarity is not required.
Catalog No. NC1801441