Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peptone from Porcine and/or Bovine Tissues, MP Biomedical

Catalog No. MP210480810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
10 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210480810 10 lb.
MP210480801 1 lb.
MP210480805 5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP210480810 Supplier MP Biomedicals, Inc Supplier No. 0210480810
Add to Cart
Add to Cart

Peptone from porcine and/or bovine is Bacteriological Grade. It is a water-soluble product and is a mixture of polypeptides and amino acids formed by the partial hydrolysis of protein.

  • Peptone is produced from porcine and/or bovine tissues
  • The tissues are digested enzymatically to create small peptides and then spray dried
  • Peptone is used in culture media in bacteriology
  • The growth requirements of most non fastidious organisms will be fulfilled by the range of amino acids, peptides, and proteases in this mixture
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
pH 5.5 to 7.0 (10% aq. soln.)(literature)
Packaging Plastic
Quantity 10 lb.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.