Learn More
Alkali Scientific Peptone, Powder, 10 Kg
SDP
Catalog No. 5000659856
Encompass_Preferred
Peptone Powder, 10 Kg is a bulk-size nutrient medium used to support the growth of a wide range of microorganisms, including bacteria, yeasts, and fungi. Peptone is derived from enzymatic protein digestion and provides essential amino acids, peptides, and other growth factors required for optimal microbial development. This 10kg package is designed for large-scale microbiological applications, including industrial fermentation, microbial culture production, and laboratory research. Peptone is crucial for manufacturing biofuels, pharmaceuticals, and enzymes, making it an indispensable component of research and production processes in biotechnology, food, and pharmaceutical industries.
Catalog No. 50-006-59856 Supplier Alkali Scientific Supplier No. EPT009CP
Peptone Powder, 10 Kg is a bulk-size nutrient medium used to support the growth of a wide range of microorganisms, including bacteria, yeasts, and fungi. Peptone is derived from enzymatic protein digestion and provides essential amino acids, peptides, and other growth factors required for optimal microbial development. This 10kg package is designed for large-scale microbiological applications, including industrial fermentation, microbial culture production, and laboratory research. Peptone is crucial for manufacturing biofuels, pharmaceuticals, and enzymes, making it an indispensable component of research and production processes in biotechnology, food, and pharmaceutical industries.