Learn More
Research Products International Corp Peptone, Powder, 10 KG
SDP
Catalog No. 501977009
Encompass_Preferred
Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Low NaCl content suitable for tissue culture.
Total Nitrogen (TN): 15.0-16.0%
Amino Nitrogen (AN): 3.0-3.5%
AN/TN Ratio: 0.21-0.23
Catalog No. 50-197-7009 Supplier Research Products International Corp Supplier No. P2024010000.0
Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Low NaCl content suitable for tissue culture.
Total Nitrogen (TN): 15.0-16.0%
Amino Nitrogen (AN): 3.0-3.5%
AN/TN Ratio: 0.21-0.23