Learn More
Alkali Scientific Peptone, Powder, 100 G
SDP
Catalog No. 5000659855
Encompass_Preferred
Peptone Powder, 100 G is a high-quality nutrient source used in the preparation of microbiological media for the cultivation of a wide range of microorganisms. Peptone is an enzymatic digest of proteins, providing essential amino acids, peptides, and other nutrients required for microbial growth. This 100g package is suitable for smaller scale media preparations and is used in various microbiological, biotechnological, and pharmaceutical applications. Peptone supports the growth of both bacteria and fungi, making it essential for applications such as fermentation studies, microbial isolation, and the production of industrial enzymes, antibiotics, and biofuels.
Catalog No. 50-006-59855 Supplier Alkali Scientific Supplier No. EPT007CP
Peptone Powder, 100 G is a high-quality nutrient source used in the preparation of microbiological media for the cultivation of a wide range of microorganisms. Peptone is an enzymatic digest of proteins, providing essential amino acids, peptides, and other nutrients required for microbial growth. This 100g package is suitable for smaller scale media preparations and is used in various microbiological, biotechnological, and pharmaceutical applications. Peptone supports the growth of both bacteria and fungi, making it essential for applications such as fermentation studies, microbial isolation, and the production of industrial enzymes, antibiotics, and biofuels.