Learn More
Alkali Scientific Peptone, Powder, 500 G
SDP
Catalog No. 5000660280
Encompass_Preferred
Peptone, Powder, is an enzymatic digest derived from microbial or animal sources and is used as a nutrient source in microbiological and biochemical applications. This 500g pack provides a high-quality peptone that supports the growth of a wide range of microorganisms, including bacteria, yeasts, and molds. It is commonly used in the preparation of growth media for microbial cultivation, including media for fermentation and enzyme production. Peptone serves as a rich source of amino acids, peptides, and other essential growth factors required by microorganisms. It is widely used in the preparation of complex media for both research and industrial applications.
Catalog No. 50-006-60280 Supplier Alkali Scientific Supplier No. EPT010CP
Peptone, Powder, is an enzymatic digest derived from microbial or animal sources and is used as a nutrient source in microbiological and biochemical applications. This 500g pack provides a high-quality peptone that supports the growth of a wide range of microorganisms, including bacteria, yeasts, and molds. It is commonly used in the preparation of growth media for microbial cultivation, including media for fermentation and enzyme production. Peptone serves as a rich source of amino acids, peptides, and other essential growth factors required by microorganisms. It is widely used in the preparation of complex media for both research and industrial applications.