Learn More
AG Scientific Inc Peptone Powder 500 G
Catalog No. 502268357
Encompass_Preferred
Peptone is a mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Low NaCl content suitable for tissue culture.
Catalog No. 50-226-8357 Supplier AG Scientific Inc Supplier No. P2841500GM
Peptone is a mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue. Low NaCl content suitable for tissue culture.