Learn More
Alkali Scientific Peptone, Soy, 1 Kg
SDP
Catalog No. 5000659854
Encompass_Preferred
Peptone, Soy, 1kg, is a plant-based enzymatic digest derived from soybeans, providing a rich nutrient source for microbial growth in both research and industrial applications. This peptone is ideal for growing a variety of microorganisms, including bacteria and fungi, that require complex organic nitrogen sources. Soy Peptone is commonly used in the preparation of fermentation media, microbiological culture media, and assays that require plant-based nutrients. It is often favored in vegetarian or vegan applications and offers an alternative to animal-derived peptones, making it suitable for research where plant-based sources are preferred or required.
Catalog No. 50-006-59854 Supplier Alkali Scientific Supplier No. EPT012CP
Peptone, Soy, 1kg, is a plant-based enzymatic digest derived from soybeans, providing a rich nutrient source for microbial growth in both research and industrial applications. This peptone is ideal for growing a variety of microorganisms, including bacteria and fungi, that require complex organic nitrogen sources. Soy Peptone is commonly used in the preparation of fermentation media, microbiological culture media, and assays that require plant-based nutrients. It is often favored in vegetarian or vegan applications and offers an alternative to animal-derived peptones, making it suitable for research where plant-based sources are preferred or required.