Learn More
Alkali Scientific Peptone, Soy, 500 G
SDP
Catalog No. 5000659779
Encompass_Preferred
Peptone, Soy, 500g, is a plant-based enzymatic digest of soybeans, providing an excellent source of organic nitrogen and growth factors for a wide range of microorganisms. This peptone is commonly used in the formulation of culture media for microbiological research and industrial applications such as fermentation, enzyme production, and microbial assays. Soy Peptone offers a sustainable and ethical alternative to animal-derived peptones, making it ideal for applications that require plant-based or vegan components. It promotes the growth of bacteria, yeasts, and fungi by providing essential amino acids and peptides necessary for cellular metabolism and reproduction.
Catalog No. 50-006-59779 Supplier Alkali Scientific Supplier No. EPT013CP
Peptone, Soy, 500g, is a plant-based enzymatic digest of soybeans, providing an excellent source of organic nitrogen and growth factors for a wide range of microorganisms. This peptone is commonly used in the formulation of culture media for microbiological research and industrial applications such as fermentation, enzyme production, and microbial assays. Soy Peptone offers a sustainable and ethical alternative to animal-derived peptones, making it ideal for applications that require plant-based or vegan components. It promotes the growth of bacteria, yeasts, and fungi by providing essential amino acids and peptides necessary for cellular metabolism and reproduction.