Learn More
Research Products International Corp Peptone, Soy, 500 Grams
SDP
Catalog No. 50213639
Encompass_Preferred
From papaic hydrolysis of soybean meal. Low NaCl content.
Catalog No. 50-213-639 Supplier Research Products International Corp Supplier No. P50300500.0
From papaic hydrolysis of soybean meal. Low NaCl content.