Learn More
Medchemexpress LLC Peptone from soya | 91079-46-8 | 500 G
SDP
Catalog No. 5000283392
Encompass_Preferred
Peptone from soya is a soy-derived enzymatic digest supplied as a solid powder. It is used as a nutrient source providing peptides, amino acids, and growth factors for microbiological media, fermentation, and biochemical assays.
  • Soy-derived enzymatic digest as a nutrient source.
  • Provides peptides, amino acids, and growth factors.
  • Suitable for microbiological media, fermentation, and biochemical assays.
  • Solid powder form, convenient for media preparation.
  • Available in 500 G packages.
  • Store at room temperature; shelf life 3 years.
  • Typical specifications: total nitrogen ≥8% and loss on drying 0-10% (example result 3.5%).
Catalog No. 50-002-83392 Supplier Medchemexpress LLC Supplier No. HYW133891500G
Peptone from soya is a soy-derived enzymatic digest supplied as a solid powder. It is used as a nutrient source providing peptides, amino acids, and growth factors for microbiological media, fermentation, and biochemical assays.
  • Soy-derived enzymatic digest as a nutrient source.
  • Provides peptides, amino acids, and growth factors.
  • Suitable for microbiological media, fermentation, and biochemical assays.
  • Solid powder form, convenient for media preparation.
  • Available in 500 G packages.
  • Store at room temperature; shelf life 3 years.
  • Typical specifications: total nitrogen ≥8% and loss on drying 0-10% (example result 3.5%).