Learn More
Medchemexpress LLC Peptone from soya | 91079-46-8 | MFCD07371265 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW1338911KG
Encompass_Preferred
Peptone from soya is an enzymatic digest of soybean proteins used as a nutrient supplement in microbiological and cell culture media. It supplies peptides, free amino acids, and other growth factors to support microbial growth and biochemical assays. Supplied as a dry powder for laboratory use.
Catalog No. 50-002-83391