Learn More
Alkali Scientific Peptone Water 0.1% 500mL Bottle, 1/Pk
SDP
Catalog No. 5000663093
Encompass_Preferred
Peptone Water 0.1% is a sterile, non selective diluent widely used in food safety testing, environmental microbiology, and water quality analysis. This 500 mL bottle format is designed for high volume laboratory workflows where larger quantities of diluent are required for sample preparation. The solution is commonly used for creating primary suspensions of solid food samples prior to microbiological testing, often by adding a measured amount of diluent to homogenized samples in sterile bags. The formulation contains peptone and sodium chloride to maintain osmotic balance and protect microorganisms from stress during handling. By preserving microbial viability without stimulating significant growth, this diluent ensures accurate results in subsequent plating, enumeration, or enrichment procedures.
Catalog No. 50-006-63093 Supplier Alkali Scientific Supplier No. PPEB00
Peptone Water 0.1% is a sterile, non selective diluent widely used in food safety testing, environmental microbiology, and water quality analysis. This 500 mL bottle format is designed for high volume laboratory workflows where larger quantities of diluent are required for sample preparation. The solution is commonly used for creating primary suspensions of solid food samples prior to microbiological testing, often by adding a measured amount of diluent to homogenized samples in sterile bags. The formulation contains peptone and sodium chloride to maintain osmotic balance and protect microorganisms from stress during handling. By preserving microbial viability without stimulating significant growth, this diluent ensures accurate results in subsequent plating, enumeration, or enrichment procedures.