Learn More
Alkali Scientific Peptone Water 0.1% 9mL Tube, 10/Pk
SDP
Catalog No. 5000663125
Encompass_Preferred
Peptone Water 0.1% is a sterile diluent commonly used in food microbiology, environmental testing, and water analysis for preparing microbial suspensions and serial dilutions. The solution contains a low concentration of peptone that helps maintain the viability of microorganisms without significantly promoting growth during the dilution process. Each tube contains exactly 9 mL of sterile diluent, allowing laboratories to easily prepare standard 1:10 serial dilutions by adding 1 mL of sample. The formulation also maintains osmotic balance, helping prevent cell damage that may occur when microorganisms are diluted in distilled water. Peptone Water 0.1% is widely used for preparing samples prior to plating, enumeration, or further microbiological analysis.
Catalog No. 50-006-63125 Supplier Alkali Scientific Supplier No. PPET00
Peptone Water 0.1% is a sterile diluent commonly used in food microbiology, environmental testing, and water analysis for preparing microbial suspensions and serial dilutions. The solution contains a low concentration of peptone that helps maintain the viability of microorganisms without significantly promoting growth during the dilution process. Each tube contains exactly 9 mL of sterile diluent, allowing laboratories to easily prepare standard 1:10 serial dilutions by adding 1 mL of sample. The formulation also maintains osmotic balance, helping prevent cell damage that may occur when microorganisms are diluted in distilled water. Peptone Water 0.1% is widely used for preparing samples prior to plating, enumeration, or further microbiological analysis.