Learn More
Research Products International Corp Peptone Water, 5 Kilograms
SDP
Catalog No. 501368492
Encompass_Preferred
Minimal growth medium for determining carbohydrated fermentation capabilities.
Catalog No. 50-136-8492 Supplier Research Products International Corp Supplier No. P203005000.0
Minimal growth medium for determining carbohydrated fermentation capabilities.