Learn More
Biolog Inc PY BROTH - Peptone Yeast Extract Broth, 10 Tubes/Pkg
SDP
Catalog No. NC0951300
Encompass
This item has a minimum qty of 15 per supplier requirements.
Peptone Yeast Extract Broth Based Media are enriched nonselective broth used for growth & identification of anaerobic bacteria based on their biochemical features. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.PY Broth contains hemin & vitamin K for the recovery of fastidious organisms such as Prevotella spp and Porphyromonas spp. PRAS (Pre-reduced anaerobically sterilized) biochemicals are considered the gold standard for biochemical testing and characterization of anaerobes. Adding specific biochemical, the Peptone Yeast Extract Broth Based Media can determine whether anaerobic bacteria will utilize the biochemical or if growth is stimulated or inhibited.
Catalog No. NC0951300 Supplier Biolog Inc Supplier No. AS821
This item has a minimum qty of 15 per supplier requirements.
Peptone Yeast Extract Broth Based Media are enriched nonselective broth used for growth & identification of anaerobic bacteria based on their biochemical features. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.PY Broth contains hemin & vitamin K for the recovery of fastidious organisms such as Prevotella spp and Porphyromonas spp. PRAS (Pre-reduced anaerobically sterilized) biochemicals are considered the gold standard for biochemical testing and characterization of anaerobes. Adding specific biochemical, the Peptone Yeast Extract Broth Based Media can determine whether anaerobic bacteria will utilize the biochemical or if growth is stimulated or inhibited.