Learn More
Thomas Scientific Prefilters, 5uM GMF, 47mm, 50/Pk.
SDP
Supplier:  Thomas Scientific 1217N32
Encompass
ValuSep 5um Glass Micro Fiber (GMF) prefilters are made of two separate layers of glass fiber in a very thin format for highly efficient capture and retention of particles that could prematurely clog an SPE filter.
Catalog No. NC0614235