Learn More
Hach Company Peracetic Acid Chemkey
Supplier:  Hach Company 8635200
Encompass
Peracetic Acid Chemkeys range 0 04 - 50 0 mg/L PAA For SL1000/SL250 Portable Parallel Analyzer (PPA)
Catalog No. 50-002-06967