Learn More
CHEMetrics™ by AquaPhoenix Peracetic Acid SAM Kit
SDP
Catalog No. 13299713
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-713 30
1 options
Catalog No. 13-299-713 Supplier CHEMetrics™ by AquaPhoenix Supplier No. I2020
Add to Cart
Add to Cart
Instrumental peracetic acid test kit using the total chlorine DPD method for PAA analysis. Produces a pink color in the presence of peracetic acid for measurement.
  • SAM Kit I-2020 comes in a plastic case and contains everything needed to perform 30 tests: Peracetic Acid Test Kit (K-7913) Vacu-vials Kit, SAM Photometer, light shield, 4 AAA batteries, screwdriver, and instructions.
Specifications
Analytes Peracetic Acid
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 5.00 ppm
Type Peracetic Acid SAM Kit