Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peracetic Acid Solution, Approx. 32%, Spectrum™ Chemical
SDP

Catalog No. 18605026 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-605-026 500 mL
18-605-025 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-605-026 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1023500ML

missing translation for 'validMainframeMsg'

P1023, C2H4O3

Spectrum™ Chemical Peracetic Acid Solution

Chemical Identifiers

CAS 79-21-0
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Max % 0.355
Quantity 500 mL
Percent Purity ∼32%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.