Learn More
CHEMetrics™ by AquaPhoenix Peracetic Acid Standard
SDP
Catalog No. 13299542
Change view
Click to view available options
Range:
1.0 ppm
Catalog No. Range
13-299-542 1.0 ppm
1 options
Catalog No. 13-299-542 Supplier CHEMetrics™ by AquaPhoenix Supplier No. A7925
Add to Cart
Add to Cart
Peracetic acid standard for verifying the accuracy and reliability of PAA measurements. Provides a convenient method for measurement confirmation.
  • CHEMetrics™ A-7925 Peracetic Acid (PAA) Standard offers the analyst an easy method to confirm the accuracy and reliability of Peracetic Acid measurements.
  • Conveniently packaged in a cardboard box, each pack contains 10 ampoules, 1 bottle of Activator Solution, a tip breaking tool, instructions, and a lot-specific Certificate of Analysis.
Specifications
pH Range 7
Volume (Metric) 0.5 mL
Range 1.0 ppm
Type Peracetic Acid Standard