Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Peracetic Acid Test Kit
SDPSCI_ED

Catalog No. S72313 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72313 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72313 Supplier LaMotte™ Supplier No. 719102
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Tests for peracetic acid and hydrogen peroxide using the dropper bottle method

  • Range/Sensitivity: 1 drop = 50ppm Peroxide, 1 drop = 15ppm Peracetic Acid
  • No. of tests: 50
  • No. of reagents: 5

Specifications

For Use With (Application) Industrial water boiling and cooling, water and wastewater
Includes 30mL sulfuric acid, 15mL ferroin indicator, 2 x 30mL hydrogen peroxide titrant, 30mL potassium iodide 20% solution, 60mL peracetic acid titrant, test tube, 5 to 10 to 25mL, plastic, with cap, 0.5mL plastic pipet
Quantity 50 Tests
Test Method Dropper bottle
Type Peracetic Acid Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.