Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Peracetic Acid Titrant (Sodium Thiosulfate, 0.1N), Lamotte™
SDPSCI_ED

Catalog No. S72333 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S72333 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S72333 Supplier LaMotte™ Supplier No. S6155H
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Test Kit Reagent

Specifications

Color Colorless
Composition 144-55-8, 10102-17-7, 7732-18-5
Description Sodium Thiosulfate, 0.1N
Format Liquid
Quantity 60 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.