Learn More
CHEMetrics™ by AquaPhoenix Peracetic Acid Water Quality Visual Test Kit
SDP
Catalog No. 13299625
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-625 30
1 options
Catalog No. 13-299-625 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K7904
Add to Cart
Add to Cart
Peracetic acid visual test kit for measuring PAA concentration in water. Comes in a plastic case and contains everything needed to perform 30 tests.
  • K-7904 features self-filling ampoules for the colorimetric analysis of the disinfectant peracetic acid (PAA) in water.
  • Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low and High Range Comparators, Activator Solution, 25 mL sample cup, and instructions.
Specifications
Analytes Peracetic Acid
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 1 and 0 to 5 ppm
Type Water Quality Visual Test Kit