Learn More
Apexbio Technology LLC Peramivir Trihydrate 1041434-82-5 10mg
SDP
Catalog No. 5000088125
Encompass_Preferred
Peramivir Trihydrate (CAS 1041434-82-5) is a small-molecule inhibitor targeting influenza viral neuraminidase It is designed to inhibit neuraminidase activity thereby impeding viral particle release and the spread of infection Peramivir Trihydrate exerts its biological activity primarily through competitive inhibition of neuraminidase as a transition-state analog In experimental studies Peramivir Trihydrate demonstrates neuraminidase inhibitory activity with an IC50 of approximately 0 09 nM Based on these pharmacological properties Peramivir Trihydrate holds research potential in investigating influenza virus replication antiviral efficacy and neuraminidase-mediated viral dissemination
Catalog No. 50-000-88125 Supplier Apexbio Technology LLC Supplier No. B214710
Peramivir Trihydrate (CAS 1041434-82-5) is a small-molecule inhibitor targeting influenza viral neuraminidase It is designed to inhibit neuraminidase activity thereby impeding viral particle release and the spread of infection Peramivir Trihydrate exerts its biological activity primarily through competitive inhibition of neuraminidase as a transition-state analog In experimental studies Peramivir Trihydrate demonstrates neuraminidase inhibitory activity with an IC50 of approximately 0 09 nM Based on these pharmacological properties Peramivir Trihydrate holds research potential in investigating influenza virus replication antiviral efficacy and neuraminidase-mediated viral dissemination