Learn More
Medchemexpress LLC Peramivir trihydrate | 1041434-82-5 | 98.3% | 382.45 | 25 MG
SDP
Catalog No. 5000385131
Encompass_Preferred
Peramivir trihydrate is a highly potent, selective, and orally active influenza virus neuraminidase (NA) inhibitor. It is intended for research use only.
  • Highly potent and selective influenza virus neuraminidase (NA) inhibitor
  • Has IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes
  • Exhibits no toxicity to macrophages at tested concentrations
  • Inhibits cytokine release in LPS-induced hPBMCs
  • Inhibits LPS-induced cytokine storm and attenuates acute lung injury in mice
  • Prolongs survival in cytokine storm syndrome model mice
Catalog No. 50-003-85131 Supplier Medchemexpress LLC Supplier No. 25MGHY17015
Peramivir trihydrate is a highly potent, selective, and orally active influenza virus neuraminidase (NA) inhibitor. It is intended for research use only.
  • Highly potent and selective influenza virus neuraminidase (NA) inhibitor
  • Has IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes
  • Exhibits no toxicity to macrophages at tested concentrations
  • Inhibits cytokine release in LPS-induced hPBMCs
  • Inhibits LPS-induced cytokine storm and attenuates acute lung injury in mice
  • Prolongs survival in cytokine storm syndrome model mice