Learn More
Medchemexpress LLC HY-17015 10mg , Peramivir (trihydrate) CAS:1041434-82-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1701510MG
Encompass_Preferred
HY-17015 10mg Peramivir (trihydrate) CAS: 1041434-82-5 Peramivir trihydrate (RWJ-270201 trihydrate;BCX-1812 trihydrate) is a highly potent, selective and orally active influenza virus neuraminidase (NA) inhibitor, with IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0202