Learn More
Selleck Chemical LLC Peramivir Trihydrate S2716-5mg
SDP
Catalog No. 503112511
Encompass_Preferred
Peramivir Trihydrate (BCX-1812 RWJ-270201 S-021812) is a trihydrate of the anti-infection agent peramivir which is a transition-state analogue and a potent specific influenza viral neuraminidase inhibitor with an IC50 of median 0 09 nM
Catalog No. 50-311-2511 Supplier Selleck Chemical LLC Supplier No. S27165MG
Peramivir Trihydrate (BCX-1812 RWJ-270201 S-021812) is a trihydrate of the anti-infection agent peramivir which is a transition-state analogue and a potent specific influenza viral neuraminidase inhibitor with an IC50 of median 0 09 nM