Learn More
Cayman Chemical Percazn 10mg
SDP
Catalog No. 502949827
Encompass_Preferred
A typical antipsychotic; binds to the dopamine D1 receptor and androgen receptor (Kis = 0.01 and 3 µM, respectively); an α1-AR antagonist (IC50 = 4.1 nM in rat forebrain homogenates); an α2-AR antagonist (IC50 = 2 µM in rat cortical homogenates); inhibits SARS-CoV-2 Mpro activity by 55% at 100 µM; increases the number of entries into, and the percentage of time spent in, the open arms of the elevated plus maze in rats at 0.075 mg/kg
Catalog No. 50-294-9827 Supplier Cayman Chemical Supplier No. 3314910 MG
A typical antipsychotic; binds to the dopamine D1 receptor and androgen receptor (Kis = 0.01 and 3 µM, respectively); an α1-AR antagonist (IC50 = 4.1 nM in rat forebrain homogenates); an α2-AR antagonist (IC50 = 2 µM in rat cortical homogenates); inhibits SARS-CoV-2 Mpro activity by 55% at 100 µM; increases the number of entries into, and the percentage of time spent in, the open arms of the elevated plus maze in rats at 0.075 mg/kg