Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Staphylococcus aureus V-8 Protease

Catalog No. PI20195 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20195 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20195 Supplier Thermo Scientific™ Supplier No. 20195
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifically cleave only at glutamic acid residues or cleave at both glutamic and aspartic residues with our immobilized Staphylococcus aureus V-8 protease kit.

Thermo Scientific™ Pierce V-8 Protease Kit contains Staphylococcus aureus V-8 protease immobilized on resin and three digestion buffers for specific cleavage at glutamic acid residues or cleavage at glutamic and aspartic acid residues.

Has two pH optima, one at pH 4.0 and the other at pH 7.8 and is specific for cleavage at the carboxy terminus of glutamic acid and aspartic acid residues. The protease is further specific for only glutamic digestion in buffers, which do not contain phosphate at either pH optimum. Phosphate buffers at pH 7.8 will facilitate digestion at both glutamic and aspartic acid residues.

Highlights:

  • Specificity for glutamic acid is achieved in ammonium bicarbonate, pH 7.8 or ammonium acetate, pH 4
  • Enzyme is active in the presence of many denaturing agents such as SDS, urea and guanidine·HCl
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Staphylococcus aureus V-8 Protease
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Form Solid
Quantity 5 mg
Product Type Protease
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.