Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Perchloric Acid

Catalog No. 3114211L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-142-11L 1 L
31-142-150ML 50 mL
31-142-1250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 31-142-11L Supplier Sigma Aldrich Supplier No. 3114211L
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Perchloric acid can be used as:A reaction medium in oxidation reactions A reagent for the preparation of fluorinated polymer electrolytes using poly (vinylidenefluoride-cohexafluoropropylene) (PVDF-HFP) for fuel cell applications A dopant in the preparation of perchloric acid-functionalized polyaniline (HClO4/PANI) catalyst for the synthesis of 2-substituted benzothiazoles by condensation reaction between o-aminothiophenol with aldehydes.

Specifications

CAS 7601-90-3
Density 1.664 g/mL at 25°C
Molecular Formula HClO4
Linear Formula HClO4
Quantity 1 L
MDL Number MFCD00011325
Synonym PCA
Molecular Weight (g/mol) 100.46 g/mol
Biological Activity Thyroid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.