Learn More
Aqua Solutions Perchloric Acid 0.1 Normal in IPA,
SDP
Catalog No. NC1639229
Encompass_Preferred
Melting point/Melting range: -89.5 °C (-129 °F), Boiling point/Boiling range: 82 °C (180 °F), Flash point: 13 °C (55 °F)
Catalog No. NC1639229 Supplier Aqua Solutions Supplier No. DC126245
Melting point/Melting range: -89.5 °C (-129 °F), Boiling point/Boiling range: 82 °C (180 °F), Flash point: 13 °C (55 °F)