Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid Greater than-or-equal-to 70% AR Select™ (ACS), Macron Fine Chemicals™

Catalog No. MK8828500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-8828-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-8828-500 Supplier Avantor Supplier No. 882814
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7601-90-3
Packaging Glass
Assay Percent Range 60%
Quantity 500 mL
Molecular Formula ClHO4
Physical Form Liquid
MDL Number MFCD00011325
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES O[Cl](=O)(=O)=O
IUPAC Name perchloric acid
Molecular Weight (g/mol) 100.45
PubChem CID 24247
ChEBI CHEBI:29221
Formula Weight 100.46
Grade ACS
Chemical Name or Material Perchloric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.