Learn More
Sigma Aldrich Fine Chemicals Biosciences Perchloric acid ACS reagen
Catalog No. 502898080
Encompass
Perchloric acid (HClO4) an oxygen acid of chlorine is strongly acidic in nature. Hydrates of perchloric acid are obtained on its disSOLUTIONon in water.
Catalog No. 50-289-8080 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2442522.5L
Perchloric acid (HClO4) an oxygen acid of chlorine is strongly acidic in nature. Hydrates of perchloric acid are obtained on its disSOLUTIONon in water.