Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, OmniTrace™, MilliporeSigma™

Catalog No. MPX03971 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX03971 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX03971 Supplier MilliporeSigma Supplier No. PX03971
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recommended Storage

Store at room temperature.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula HClO4

Specifications

Melting Point -18°C
Boiling Point 198.7°C (1013hPa)
Packaging Glass Bottle
Quantity 500 mL
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.