Learn More
Sigma-Aldrich Percoll™ Density Gradient Medium
Catalog No. 01258898 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-898 25 mL
01-258-900 100 mL
01-258-901 1 L
01-258-974 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-258-898 Supplier Sigma-Aldrich Supplier No. P164425ML
Only null left
Add to Cart
Add to Cart
A classic medium for density gradient centrifugation of cells, viruses, and subcellular particles.
Percoll™ has been used in sperm preparation to study the energy requirements of bovine embryos.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 mL
pH 8.5 to 9.5
Product Type Density Gradient Medium