Learn More
Sigma Aldrich Fine Chemicals Biosciences PercollR pH 8.5 9.5 20 C1L
Catalog No. 502914229
Encompass
Percoll(R) is a classic medium for density gradient centrifugation of cells viruses and subcellular particles. Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). The PVP coating renders the product completely non-toxic and ideal for use with biological materials.
Catalog No. 50-291-4229 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P16441L
Percoll(R) is a classic medium for density gradient centrifugation of cells viruses and subcellular particles. Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). The PVP coating renders the product completely non-toxic and ideal for use with biological materials.