Learn More
Sigma Aldrich Fine Chemicals Biosciences PercollR pH 8.5 9.5 20 C500ML
Catalog No. 502904225
Encompass
Percoll(R) is a classic medium for density gradient centrifugation of cells viruses and subcellular particles. Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). The PVP coating renders the product completely non-toxic and ideal for use with biological materials.
Catalog No. 50-290-4225 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P1644500ML
Percoll(R) is a classic medium for density gradient centrifugation of cells viruses and subcellular particles. Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). The PVP coating renders the product completely non-toxic and ideal for use with biological materials.