Learn More
Biolegend PerCP anti-human CD3
Supplier:  Biolegend 317338
Encompass_Preferred
PerCP anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-022