Learn More
Biolegend PerCP anti-human CD45
Supplier:  Biolegend 982318
Encompass_Preferred
PerCP anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-207-5543