Learn More
Biolegend PerCP anti-human CD8a
Supplier:  Biolegend 300922
Encompass_Preferred
PerCP anti-human CD8a [HIT8a]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Marmoset; Apps: FC; Size: 100 tests
Catalog No. 50-165-639