Learn More
Biolegend PerCP anti-human TCR Vδ2
Supplier:  Biolegend 331410
Encompass_Preferred
PerCP anti-human TCR Vδ2 [B6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactive: Chimpanzee, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-167-649