Learn More
Biolegend PerCP anti-human CD19
Supplier:  Biolegend 392509
Encompass_Preferred
PerCP anti-human CD19 [4G7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8918