Learn More
Cytek Biosciences Inc Antibodies PerCP Anti-Human CD2 (RPA-2.10) ..... Cat. No. 67-0029-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 670029T025
Encompass_Preferred
Antibodies PerCP Anti-Human CD2 (RPA-2.10) ..... Cat. No. 67-0029-T025 ..... Size 25 testsThe RPA-2.10 antibody reacts with human CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD58 in the human and is expressed on the cell surface of 80-90% of human peripheral blood lymphocytes, a subset of NK cells, and all mature T cells. CD2 mediates lymphocyte adhesion and is involved in T cell activation.RPA-2.10 is reported to block mixed lymphocyte reaction. Please choose the appropriate format for each application.
Catalog No. 50-210-3609