Learn More
Cytek Biosciences Inc Antibodies PerCP Anti-Human CD5 (L17F12) ..... Cat. No. 67-0058-T500 ..... Size 500 tests
Supplier:  Cytek Biosciences Inc 670058T500
Encompass_Preferred
Antibodies PerCP Anti-Human CD5 (L17F12) ..... Cat. No. 67-0058-T500 ..... Size 500 testsThe L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is a member of the scavenger receptor superfamily and is present on approximately 70% of normal peripheral blood lymphocytes. CD5 is involved in modulating antigen receptor signaling in both T and B cells and acts through binding of CD72, its receptor expressed on B cells.
Catalog No. 50-210-3618