Learn More
Biolegend PerCP anti-mouse CD45.1
Supplier:  Biolegend 110725
Encompass_Preferred
PerCP anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-429