Learn More
Biolegend PerCP anti-mouse CD5
Supplier:  Biolegend 100616
Encompass_Preferred
PerCP anti-mouse CD5 [53-7.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-473