Learn More
Biolegend PerCP anti-mouse Ly-6G
Supplier:  Biolegend 127654
Encompass_Preferred
PerCP anti-mouse Ly-6G [1A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-112-9954