Learn More
Biolegend PerCP/Cyanine5.5 anti-human TCR Vα24-Jα18 (iNKT cell)
Supplier:  Biolegend 342914
Encompass_Preferred
PerCP/Cyanine5.5 anti-human TCR Vα24-Jα18 (iNKT cell) [6B11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactivity: Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 100 tests
Catalog No. 50-207-6536