Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD273 (B7-DC, PD-L2) [TY25]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Supplier:  Biolegend 107217
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD273 (B7-DC, PD-L2) [TY25]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-237-8663